ICS-6000 DC Column Compartment not heating
This article addresses the issue of the ICS-6000 DC column compartment not heating up. It provides a step-by-step resolution process, including checking for obstructions, inspecting fans and motors, examining the door mechanism, verifying compartment seals, and checking for loose components. If the issue persists, it advises contacting technical support for a potential replacement of the Column Compartment Fan (P/N 062226).
Issue
ICS-6000 DC Compartment not heating up.
Environment:
- Product: ICS-6000 DC
- Instrument: ICS-6000
Resolution:
- Check for Obstructions: Ensure that there are no obstructions in the compartment.
- Inspect Fans and Motors: Verify that the fans and motors inside the compartment are functioning correctly.
- Examine Door Mechanism: Check the door mechanism to ensure it is operating smoothly.
- Verify Compartment Seals: Ensure that all seals and gaskets around the compartment door are intact.
- Check for Loose Components: Look for any loose components inside the compartment that can limit temperature performance.
- Contact Technical Support: If the issue persists after these checks, replacement may be needed: Column Compartment Fan (P/N 062226)