IC Suppressor Leaking
Issue:
IC suppressor has a high external back pressure and is leaking.
Environment:
IC Suppressor
Resolution:
- Monitor the back pressure system.
- Look for an increase in pressure over time.
- Remove any blockages.
- Clean spill.
- Locate and remove upstream/downstream source of blockage.
Cause:
Bad Regen out connection