IC Tubing
Issue
How to correctly seat ferrule onto IC tubing before fitting to avoid dead volume.
Environment
IC Instruments
Resolution
When connecting tubing to a port, make sure the 10-32 ferrule and fitting bolt are at least 2 mm (0.1 in) from the end of the tubing before you insert the tubing into the port. Do not position the ferrule and fitting bolt flush with the end of the tubing.
Analysis
Cause
Attachment(s)
| File | Last Modified |
|---|---|
| Tubing Cut.png | September 25, 2023 |
| Ferrule seating.PNG | September 25, 2023 |