Instrument unable to control temperature
Issue
Unable to control temperature
Environment
Dionex ICS-4000 Capillary HPIC System
Resolution
- Verify that all installed temperature control devices are at their set temperatures and are stable.
- When the temperature zone is off, the background is red.
- When the temperature zone is on and the set temperature has not yet been reached, the background is amber.
- When the temperature zone is on and the set temperature has been reached, the background is green. Record any discrepancies.
Note: The temperature of the column heater must be set to at least 2 °C above the compartment temperature. The temperature of the CD cell heater must be set to at least 7 °C above the compartment temperature.
If this is good and there is an error message in the audit trail. Refer to the operator manual.