Rise in baseline during a gradient
Issue
I am using KOH RFIC with an application that has a gradient, but I am seeing large baseline rises during each run, what could be causing it?
Environment
Ion Chromatography
Resolution
CR-ATC Cleanup:
The CR-ATC for normal day-to-day operation does not require a cleanup. However, if the CR-ATC is exposed accidentally to high levels of anionic contaminants or is converted to the -OH form to other anionic forms such as carbonate, the device requires a cleanup using 2.0 M NaOH.
- Disconnect all lines to the CR-ATC.
- Connect a line from the Eluent In port to the Regen In port on the CR-ATC. Direct the Regen Out port to waste.
- Prepare a fresh solution of 2.0 M NaOH from a 50% w/w NaOH solution.
- Use the trap column/suppressor clean-up kit (P/N 059659) to deliver 100 mL of 2.0 M NaOH solution through the Eluent Out port of the CR-ATC column, or 10 mL to a CR-ATC capillary column.
- Rinse the CR-ATC unit with 10 mL (1 mL for CR-ATC capillary columns) DI water before plumbing it back to the system.
Cause
The Dionex CR-TC is most likely contaminated. Clean the trap column as instructed in the Dionex CR-TC manual. The manual is provided on the Thermo Scientific Reference Library DVD (P/N 053891).