System does not power on
Issue
No power.
Environment
Integrion
Resolution
- Unplug and re-seat the plug on the rear of the system. Does it power on?
- Try another power source, does it power on?
- Change the main power fuses, per the Dionex Integrion HPIC System Operator’s Manual. Does it power on?
- If steps 1-3 have been performed and the system still does not power on, contact Technical Support for Dionex products.
In the U.S. and Canada, call 1-800-532-4752. Outside the U.S. and Canada, call the nearest Thermo Scientific office.