USB Cannot Connect to System
Issue
- USB cannot connect to system
Environment
- ICS-2100
Resolution
-
Reboot system. Check if communication is established.
-
Check USB cable and check if communication is established.
-
Restore computer back to restore point before USB connection issue occurred. System should communicate successfully.
-
Perform test injections to determine if the system is operating as intended.