Error Code 52 - Module software incomplete
Issue
Vanquish pump experiencing error Code 52: Module software incomplete.
Environment
- Vanquish Core
- VC-A12-A-02
- VC-A13-A-02
- Vanquish Flex
- VF-A10-A-02
- VF-A40-A-02
- Vanquish Horizon
- VH-A10-A-02
- VH-A40-A-02
- Vanquish Pump
Resolution
Verify that the correct firmware file was selected. (Vanquish pump Firmware files)
Repeat the download.
Cause
Firmware failure.