Error Code 89 - Liquid leak sensor missing or defective
Issue
Vanquish pump experiencing error Code 89: Liquid leak sensor missing or defective
Environment
- Vanquish Core
- VC-A12-A-02
- VC-A13-A-02
- Vanquish Flex
- VF-A10-A-02
- VF-A40-A-02
- Vanquish Horizon
- VH-A10-A-02
- VH-A40-A-02
- Chromeleon
- Vanquish Pump
Resolution
- Disable the leak sensor functionality in the Chromeleon software by setting Leak Sensor Mode to Disabled.
- Contact Thermo Fisher Scientific Technical Support for assistance.
Cause
Hardware failure.