Thermo Fisher Scientific
Error Code 90 - Download firmware mismatch
Issue
Vanquish Autosampler experiencing Error Code 90: Download firmware mismatch
Environment
  • Vanquish Core
    • VC-A12-A-02
    • VC-A13-A-02
  • Vanquish Flex
    • VF-A10-A
    • VF-A10-A-02
    • VF-A40-A-02
  • Vanquish Horizon
    • VH-A10-A-02
    • VH-A40-A-02
Resolution
If required, repeat the download with a firmware version later than the version currently installed in the device.
Cause
Firmware failure.
  • Was this article helpful?