X tag not detected
Issue
- x tag not detected.
- x = detector component that could not be detected.
Environment
- Vanquish Core
- VC-D11
- VC-D12
- Vanquish Flex
- VF-D11
- Chromeleon
Resolution
- Open and close the front door.
- Disconnect and connect the detector in Chromeleon.
- If the error persists, turn off the module.
- Wait for 5 seconds and turn on the module again.
- Check the flow cell.
Cause
Instrument not ready.