Temperature setpoint cannot be achieved
Issue
Unit is experiencing temperature setpoint that cannot be achieved.
Environment
- Vanquish Core
- VC-A12
- VC-A13
- Vanquis Flex
- VF-A10
- VF-A40
- Vanquish Horizon
- VH-A10
- VH-A40
Resolution
- Check if the door is closed properly.
- If the Charger was removed from the Charger opening, close the Charger opening on the autosampler properly with foam part and metal sheet cover.
Cause
- The ambient temperatures may be out of specified range.
- The sample compartment may not be insulated sufficiently.