Thermo Fisher Scientific
Status Screen
Issue
If the self test was successful, the initial screen changes to the status screen.
Environment
UltiMate 3000 Autosamplers
Resolution
The status screen shows the following information:
  • Sample position
  • Injection volume
NOTE: Autosamplers with sample thermostatting option only will display 'Actual temperature' and 'Target temperature' (in brackets) for sample thermostatting.
  • Valve position (Loop: Inject or Load) user can adapt the screen brightness and contrast to your requirements if necessary.
clipboard_e41ff7de50060f529dc252a9a9a9047ae.png
  • Was this article helpful?