Removing Reagent Rack
Issue
Removing reagent rack
Environment
- Gallery/Gallery Plus/Beermaster Analyzer 7.0
- Gallery/Gallery Plus/Beermaster Analyzer 6.0
Resolution
To remove a rack reagent:
- Select F3 > REAGENT
- Select the rack to be removed.
- Click 'remove rack.'
- When the light is on, open the cover.
- Remove the rack.
- Close the cover.