Thermo Fisher Scientific
How to remove a reagent rack
Issue
How do I remove a reagent rack?
Environment
  • Gallery
  • Gallery Plus
  • Beermaster Analyzer 6.0
  • Beermaster Analyzer 7.0
Resolution
To remove a rack reagent:
  1. Select F3 > REAGENT 
  2. Select the rack to be removed. 
  3. Click 'remove rack.'
  4. When the light is on, open the cover.
  5. Remove the rack.
  6. Close the cover.
  • Was this article helpful?