Thermo Fisher Scientific
How to check reagent barcodes
Issue
How do I check reagent barcodes?
Environment
  • Gallery
  • Gallery Plus
  • Beermaster Analyzer 6.0
  • Beermaster Analyzer 7.0
Resolution
To check the content of the reagent rack:
1. Remove the rack from the analyzer. 
2. Ensure the barcodes are properly aligned. 
3. Place the rack back on the analyzer. 
NOTE: The reagents' barcodes are read and the view is updated.  
  • Was this article helpful?