Thermo Fisher Scientific
How to reduce the risk of corrosion
Issue
How do I reduce the risk of corrosion? 
Environment
  • Gallery
  • Gallery Plus
  • Beermaster Analyzer 6.0
  • Beermaster Analyzer 7.0
Resolution
NOTE: Special attention should be paid for the daily maintenance of the analyzer. 
Daily Maintenance:
  • Empty the cuvette waste bin of used cuvettes preferably after each test run or at least at the end of the day while the analyzer is still on.
  • Flush the cuvette waste bin with water and remove any residue.
  • At the end of the day, take the reagents away from the analyzer. 
  • Open the analyzer's main vent to ventilate the analyzer and leave the cover open for at least 30 minutes.
  • Wipe all visible reagent spills with a cloth moistened with mild detergent. 
  • Empty the waste water container and clean with tap water. 
BE ADVISED: Ensure that the laboratory is adequately and properly ventilated. 
  • Was this article helpful?