Correct way to adjust the Vacuum Safety and Pressure Switch
To adjust the vacuum safety and pressure switch on Model 1025, remove the right end panel to locate the switches. The vacuum switch is on the left, and the pressure switch with a capacitor attached to the micro switch is on the right. Adjust the vacuum switch by turning the screw fully counterclockwise, then clockwise 2 ½ turns. Adjust the pressure switch by turning the screw fully counterclockwise.
Issue
Correct way to adjust the vacuum safety and pressure switch
Environment
Anaerobic System Model 1025
Resolution
Refer to the attached document for instructions.
Attachment(s)
| File | Last Modified |
|---|---|
| Correct way to adjust the Vacuum Safety and Pressure Switch.pdf | January 13, 2022 |