Thermo Fisher Scientific
Excessive mixed gas usage
To address excessive mixed gas usage in a Model 1025 Anaerobic System, ensure the supply tank pressure is between 10 to 15 psi. Verify that the glove chamber relief bubbler contains mineral oil and that the vent tube is submerged by 3/8” to 1/4”. Inspect all supply lines and the in-line filter for leaks using soapy water and a sponge brush. Additionally, check the vinyl front and gloves for leaks by opening the manual fill valve until the front balloons out, then listen or feel for leaks.
Issue
What to check for excessive mixed gas usage
Environment
Anaerobic System Model 1025
Resolution
What to check for excessive mixed gas usage on a model# 1025:
  • Check supply tank pressure, should be 10 to 15 psi
  • Make sure the glove chamber relief bubbler has mineral oil in the bottle
  • Make sure the vent tube is in the mineral oil by 3/8” to 1/4”
  • Check all supply lines and in-line filter for leaks by using soupy water and a small sponge brush
  • Check the vinyl front and gloves for leaks by opening the manual fill valve until the front balloons out, than listen or feel for leaks.
Refer to the attached document for an illustration.
  • Was this article helpful?