7033110 - Forma Model 3110 3210 Series - Operating and Maintenance Manual
The document "7033110 - Forma Model 3110, 3210 Series - Operating and Maintenance Manual" provides detailed operating and maintenance instructions for the Forma 3110 Series Single Chamber Water Jacket Incubator and the Forma 3210 Series Dual Chamber Water Jacket Incubator. It includes model numbers, CO2 sensor types (thermal conductivity (TC) and infrared (IR)), oxygen options, and voltage specifications (115V and 230V) for each model. Dual chamber models can be configured as stacked or side-by-
Description:
Operating and maintenance manual
Environment:
- Forma 3110 Series, Single Chamber Water Jacket Incubator
- Forma 3210 Series, Dual Chamber Water Jacket Incubator
Model numbers:
| Single Chamber Models | |||
| Model | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
| Dual Chamber Models | |||
| Model* | CO2 Sensor** | O2 | Voltage*** |
|
TC | No | 115 |
|
TC | No | 230 |
|
IR | No | 115 |
|
IR | No | 230 |
|
TC | Yes | 115 |
|
TC | Yes | 230 |
|
IR | Yes | 115 |
|
IR | Yes | 230 |
*The models listed as dual chamber models are stacked or dual chamber units. These units arrive as single stand-alone units. They may be stacked one on top of the other or configured side by side with opposite door swing.
**TC is a thermal conductivity sensor. IR is an infrared sensor.
***All units are 50-60 Hz.