Temperature Set or Reset Value
Issue
How do I set the Temperature value?
Environment
- Heracell VIOS 160i CR
- Heracell VIOS 250i CR
Resolution
-
Refer to the Heracell VIOS 160i/250i CR Operating Instructions for more information.
Image 1: Temperature Display
- Press theTemperature key. The Temperature menu is displayed.
- To exit the Temperature menu, press the End key.
- To enter the Temperature set value, press the Set value key.
- To increase the set value, press the + key.
- To reduce the set value, press the - key.
Image 2: Setting the Temperature Value
- To accept and save the set value, press the Enter key. The system returns to the main menu, and the temperature display shows the actual value currently measured in the workspace.