Thermo Fisher Scientific
Carbon Dioxide (CO2) Set Value
To set the CO2 value on Heracell VIOS 160i CR and Heracell VIOS 250i CR incubators, follow the specific steps provided in the user manual for these models. The manual will guide you through accessing the control panel and adjusting the CO2 settings to the desired value.
Issue 
How do I set the CO2 value?
Environment
  • Heracell VIOS 160i CR
  • Heracell VIOS 250i CR
Resolution
Refer to the Heracell VIOS 160i/250i CR Operating Instructions for more information.
Image 1: CO2 Display and CO2 Menu 
  1. Press the CO2 display key. The CO2 menu is displayed.
  2. To exit the CO2 menu, press the End key.
  3. To enter the CO2 set value, press the Set value key.
    1. To increase the set value, press the + key.
    2. To reduce the set value, press the - key.
  4. To accept and save the set value, press the Enter key. The system returns to the main menu, and the CO2 display shows the actual value currently measured in the workspace. 
  5. Deactivate the CO2 control loop by changing the set value to 0%
NOTE: 
  • Error monitoring is enabled when the control loop is deactivated.
  • Ventilate the inner chamber after changing the CO2 setpoints so that no alarm occurs..
  • Was this article helpful?