Temperature Set or Reset Value
The document titled "Temperature Set or Reset Value" provides instructions for setting the temperature value on the Heracell VIOS 160i CR and 250i CR CO2 Incubators. It explains how to access the Temperature menu, adjust the temperature set value using the + and - keys, and save the changes by pressing the Enter key. The system then returns to the main menu, displaying the actual temperature value measured in the workspace. For detailed guidance, users are referred to the Heracell VIOS 160i/250i
Issue
How do I set the Temperature value?
Environment
- Heracell VIOS 160i CR
- Heracell VIOS 250i CR
Resolution
-
Refer to the Heracell VIOS 160i/250i CR Operating Instructions for more information.
Image 1: Temperature Display
- Press theTemperature key. The Temperature menu is displayed.
- To exit the Temperature menu, press the End key.
- To enter the Temperature set value, press the Set value key.
- To increase the set value, press the + key.
- To reduce the set value, press the - key.
Image 2: Setting the Temperature Value
- To accept and save the set value, press the Enter key. The system returns to the main menu, and the temperature display shows the actual value currently measured in the workspace.