Ice formation
Issue
Formation of ice
Environment
Multifuge X Pro-Megafuge ST Plus Centrifuge Series:
| Laboratory Use Centrifuges | In Vitro Diagnostic Centrifuges | ||
| Catalog No. | Benchtop Centrifuge | Catalog No. | Benchtop Centrifuge |
| 75009900 | Multifuge X4 Pro 208-240 V ±10%, 50 / 60 Hz | 75009500 | Multifuge X4 Pro-MD 208-240 V ±10%, 50 / 60 Hz |
| 75009901 | Multifuge X4 Pro 120 V ±10%, 60 Hz | 75009501 | Multifuge X4 Pro-MD 120 V ±10%, 60 Hz |
| 75009915 | Multifuge X4R Pro 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz | 75009515 | Multifuge X4R Pro-MD 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz |
| 75009815 | Multifuge X4R Pro 220 V ±10%, 60 Hz | 75009615 | Multifuge X4R Pro-MD 220 V ±10%, 60 Hz |
| 75009916 | Multifuge X4R Pro 120 V ±10%, 60 Hz | 75009516 | Multifuge X4R Pro-MD 120 V ±10%, 60 Hz |
| 75009903 | Megafuge ST4 Plus 208-240 V ±10%, 50 / 60 Hz | 75009503 | Megafuge ST4 Plus-MD 208-240 V ±10%, 50 / 60 Hz |
| 75009904 | Megafuge ST4 Plus 120 V ±10%, 60 Hz | 75009504 | Megafuge ST4 Plus-MD 120 V ±10%, 60 Hz |
| 75009918 | Megafuge ST4R Plus 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz | 75009518 | Megafuge ST4R Plus-MD 220-240 V ±10%, 50 Hz / 230 V ±10%, 60 Hz |
Resolution
To remove the ice from the centrifugation chamber, proceed as follows:
- Open the centrifuge door.
- Remove the rotor. Let the ice melt.
- Remove the water from the centrifugation chamber.
- Clean the centrifuge chamber.
NOTE: Do not use any sharp tools, aggressive liquids or fire to fasten the melting process. If necessary use warm water to speed up the melting process.
Cause
Warm humid air in combination with a cold centrifugation chamber can lead to formation of ice.