Thermo Fisher Scientific
Storage of centrifuges
Issue
Short- and long-term storage of centrifuges
Environment
Centrifuges
Resolution
Floor Model Centrifuge
First, remove the rotor from the centrifuge and completely wipe it clean before storing it. Failure to removing the rotor can cause it to become stuck on the motor shaft and potentially may require parts replacement.
If the centrifuge is not going to be put in service for a few weeks and does not require relocation, our recommendation is to switch power off to the unit, unplug it from the wall outlet and nicely place the power cord inside of the chamber after wiping it off dry. Leave the centrifuge’s lid open.
If relocation of the centrifuge is necessary, due to the complexity of the process involved, contact Thermo Fisher (1-800-438-4851) and request to have a technician come out to assist or have in-house technician speak with technical support. Follow the same process for putting the instrument back into service.
Tabletop or Benchtop Centrifuge
First, remove the rotor from the centrifuge and completely wipe it clean before storing it. Failure to removing the rotor can cause it to become stuck on the motor shaft and potentially may require parts replacement.
Switch off the power switch of the centrifuge, unplug it from the wall outlet and nicely place the power cord in the chamber after wiping it off dry. Leave the centrifuge’s lid open.
Long Term Storage
If long term storage is necessary, make sure to decontaminate/disinfect the centrifuge and its accessories by following laoratory protocols or, if not sure, speak with Thermo Fisher’s Pre-Sales applications for guidance (866-984-3766). Avoid storing the centrifuge in direct sunlight.
  • Was this article helpful?