327451H01 - Plasma Blood Packet Layout for 51.1 Cu.Ft Drawing
This document provides blood packet layout for a 51.1 cubic feet unit. with details on the storage capacity and layout for a 250ML plasma bag shelf system, specifically referencing shelf #34475G03. Each shelf can hold 76 bags, with a total storage capacity of 1,064 bags per unit over 14 shelves.
Description:
Instruction sheet and drawing for plasma blood packet layout
Environment:
Blood Plasma 51.1 CU.FT unit:
- FFPF5030
- JPL5030
- UFP5030
Attachment(s):
| File | Last Modified |
|---|---|
| 327451H01 - Plasma Blood Packet Layout for 51.1 Cu.Ft Drawing.pdf | April 24, 2024 |