33530H37 - NGC2 High Performance Lab Refrigerators and Freezers Wiring Diagram
This document is a wiring diagram for the High Performance Laboratory Refrigerator and Freezer (HPLRF) NGC2 boards schematic. It details the connections and components such as fan motors, compressors, heaters, defrost mechanisms, and power supply boards. The diagram includes color-coded wiring instructions, voltage specifications, and component connections for various parts like evaporators, defrost drain pans, odor heaters, and frame heaters, with notes on input details and relay operations.
Description:
Electrical schematic or wiring diagram
Environment:
NGC2 HPLRF, all brands, all voltage refrigerator and freezers:
4' -12' - 23' - 30' - 45' - 50' - 75'
| UEN2320 | RFMS2320 | IGL2023 |
| UEN3020 | PPL1230 | JPL1230 |
| UFP1230 | PPL2330 | JPL2330 |
| UFP2330 | PPL3030 | JPL3030 |
| UFP3030 | LF2330 | JPL430 |
| UFP430 | LF3030 | JPL5030 |
| UFP5030 | LF430 | IEN2023 |
| UGL2320 | LF5030 | IEN2030 |
| UGL3020 | IUF3012 | IGL2023 |
| ULT1230 | IUF3023 | IGL2030 |
| ULT2330 | IUF3030 | FFMS2320 |
| ULT3030 | IUF3050 | FFPF1230 |
| ULT430 | JLF1230 | FFPF2330 |
| ULT5030 | JLF2330 | FFPF3030 |
| UFP1230 | JLF3030 | FFPF430 |
| UPF2330 | JLF430 | FFPF5030 |
| UPF3030 | JLF5030 | FFGL430 |
| UPF430 | LF1230 | FFGL2320 |
| UPF5030 | FFGL5030 | FFGL2330 |
| FFEN2320 | FFEN3020 | FFGL3020 |
| TSFMS2320 | FFGL1230 | FFGL3030 |
| PPL5030 |
Attachment(s):
| File | Last Modified |
|---|---|
| 33530H37 - Rev A - NGC2 High Performance Lab Refrigerators and Freezers Wiring Diagram.pdf | April 24, 2024 |