MC20SS-SAEE - Combo Refrigerator FreezerĀ 20CuFT Specification Drawing
This document outlines Combo refrigerator/freezer unit specifications with drawing. It has a stainless-steel exterior, aluminum interior, and dimensions of 26" width, 35.5" depth, and 82.25" height. Features include a solid stainless-steel door with key lock, four adjustable wire shelves, stem mount casters, digital controls (+4°C refrigerator, -20°C freezer), automatic defrost, and 115V 60Hz power.
Description:
Drawing and specifications. Exterior measurements dimensions.
Environment:
MC20SS-SAEE Combo Refrigerator and Freezer 20CuFT
Attachment(s):
| File | Last Modified |
|---|---|
| MC20SS-SAEE - Combo Refrigerator Freezer 20CuFT Specification Drawing.pdf | April 24, 2024 |