MF25SS-SAEE- Freezer Solid SS Single Door 25CuFT Specification Drawing
This document provides the general specifications for a stainless steel freezer. Key details include its exterior and interior dimensions, finishes, insulation type, magnetic gasket, and solid stainless steel door with a key lock. It features three adjustable epoxy-coated wire shelves, digital controller preset to -30°C, automatic defrost, 3/4 HP compressor, and uses R404A refrigerant. The unit operates at 115V with a temperature range of -30°C to -12°C, has a capacity of 24 cubic feet.
Description:
Drawing and specifications. Unit exterior measurements dimensions.
Environment:
Freezer Solid SS Single Door 25CuFT
Attachment(s):
| File | Last Modified |
|---|---|
| MF25SS-SAEE - Rev A - Freezer Solid SS Single Door 25CuFT - Drawing and Specifications.pdf | December 09, 2021 |