Err on display
This article provides troubleshooting guidance for resolving the "Err" error code displayed on various TSX model laboratory refrigerators and freezers. The error indicates an upper bottle probe failure across all models, including TSX Laboratory Refrigerators, TSX Blood Bank Refrigerators, TSX -30° Lab Freezers, TSX -30° Plasma Freezers, and TSX -20° Laboratory and Enzyme Freezers. The recommended resolution is to check for a loose probe connector and replace the upper bottle probe if necessary.
Issue
Err on display
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | ||||
| TSX2305P | V/Y/Z | A/D | ||||
| TSX2305C | V/Y/Z | A/D | ||||
| TSX3005G | V/Y/Z | A/D | ||||
| TSX3005S | V/Y/Z | A/D | ||||
| TSX3005P | V/Y/Z | A/D | ||||
| TSX3005C | V/Y/Z | A/D | ||||
| TSX4505G | V/Y/Z | A/D | ||||
| TSX4505C | V/Y/Z | A/D | ||||
| TSX5005G | V/Y/Z | A/D | ||||
| TSX5005S | V/Y/Z | A/D | ||||
| TSX5005P | V/Y/Z | A/D | ||||
| TSX5005C | V/Y/Z | A/D |
Resolution
| Err on display | ||
| Error Code | Model Number | Resolution |
| Err | TSX Laboratory Refrigerators | Upper Bottle Probe Failure: Check for loose probe connector. Replace upper bottle probe. |
| Err | TSX Blood Bank Refrigerators | Upper Bottle Probe Failure: Check for loose probe connector. Replace upper bottle probe. |
| Err | TSX -30° Lab Freezers | Upper Bottle Probe Failure: Check for loose probe connector. Replace upper bottle probe. |
| Err | TSX -30° Plasma Freezers | Upper Bottle Probe Failure: Check for loose probe connector. Replace upper bottle probe. |
| Err | TSX -20° Laboratory and Enzyme Freezers | Upper Bottle Probe Failure: Check for loose probe connector. Replace upper bottle probe. |