Error code E01 on display
This article addresses the resolution of error code E01 for various TSX refrigerator and freezer models. E01 signifies an invalid algorithm and advises checking model type settings in service mode.
Issue
Error code E01 on display
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | ||||
| TSX2305P | V/Y/Z | A/D | ||||
| TSX2305C | V/Y/Z | A/D | ||||
| TSX3005G | V/Y/Z | A/D | ||||
| TSX3005S | V/Y/Z | A/D | ||||
| TSX3005P | V/Y/Z | A/D | ||||
| TSX3005C | V/Y/Z | A/D | ||||
| TSX4505G | V/Y/Z | A/D | ||||
| TSX4505C | V/Y/Z | A/D | ||||
| TSX5005G | V/Y/Z | A/D | ||||
| TSX5005S | V/Y/Z | A/D | ||||
| TSX5005P | V/Y/Z | A/D | ||||
| TSX5005C | V/Y/Z | A/D |
Resolution
| Error code E01 on display | ||
| Error Code | Model Number | Resolution |
| E01 | TSX Laboratory Refrigerators | Communication error: Call customer service. |
| E01 | TSX Blood Bank Refrigerators | Invalid Algorithm: Check to ensure the model type is set correctly in service mode. |
| E01 | TSX -30° Lab Freezers | Invalid Algorithm: Check to ensure the model type is set correctly in service mode |
| E01 | TSX -30° Plasma Freezers | Invalid Algorithm: Check to ensure the model type is set correctly in service mode. |
| E01 | TSX -20° Laboratory and Enzyme Freezers | Invalid Algorithm: Check to ensure the model type is set correctly in service mode. |