Error code E02 on display
Error code E02 on TSX High Performance Laboratory Refrigerators and Freezers indicates a control probe failure across various models, including laboratory refrigerators, blood bank refrigerators, lab freezers, plasma freezers, and enzyme freezers. The recommended action is to check for a loose probe connector and replace the control probe if necessary. This ensures proper functioning and temperature control in critical laboratory and medical storage equipment.
Issue
Error code E02 on display
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | ||||
| TSX2305P | V/Y/Z | A/D | ||||
| TSX2305C | V/Y/Z | A/D | ||||
| TSX3005G | V/Y/Z | A/D | ||||
| TSX3005S | V/Y/Z | A/D | ||||
| TSX3005P | V/Y/Z | A/D | ||||
| TSX3005C | V/Y/Z | A/D | ||||
| TSX4505G | V/Y/Z | A/D | ||||
| TSX4505C | V/Y/Z | A/D | ||||
| TSX5005G | V/Y/Z | A/D | ||||
| TSX5005S | V/Y/Z | A/D | ||||
| TSX5005P | V/Y/Z | A/D | ||||
| TSX5005C | V/Y/Z | A/D |
Resolution
| Error code E02 on display | ||
| Error Code | Model Number | Resolution |
| E02 | TSX Laboratory Refrigerators | Control Probe Failure: Check for loose probe connector. Replace control probe. |
| E02 | TSX Blood Bank Refrigerators | Control Probe Failure: Check for loose probe connector. Replace control probe. |
| E02 | TSX -30° Lab Freezers | Control Probe Failure: Check for loose probe connector. Replace control probe. |
| E02 | TSX -30° Plasma Freezers | Control Probe Failure: Check for loose probe connector. Replace control probe. |
| E02 | TSX -20° Laboratory and Enzyme Freezers | Control Probe Failure: Check for loose probe connector. Replace control probe. |