Error code E03 on display
This article addresses the error code E03 displayed on TSX High Performance Laboratory Refrigerators and Freezers. The error signifies a defrost probe failure across various models including TSX Laboratory Refrigerators, TSX Blood Bank Refrigerators, TSX -30° Lab Freezers, and TSX -30° Plasma Freezers. The recommended resolution for this issue involves checking for a loose probe connector and replacing the defrost probe if necessary.
Issue
Error code E03 on display
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | |||
| TSX2305P | V/Y/Z | A/D | |||
| TSX2305C | V/Y/Z | A/D | |||
| TSX3005G | V/Y/Z | A/D | |||
| TSX3005S | V/Y/Z | A/D | |||
| TSX3005P | V/Y/Z | A/D | |||
| TSX3005C | V/Y/Z | A/D | |||
| TSX4505G | V/Y/Z | A/D | |||
| TSX4505C | V/Y/Z | A/D | |||
| TSX5005G | V/Y/Z | A/D | |||
| TSX5005S | V/Y/Z | A/D | |||
| TSX5005P | V/Y/Z | A/D | |||
| TSX5005C | V/Y/Z | A/D |
Resolution
| Error code E03 on display | ||
| Error Code | Model Number | Resolution |
| E03 | TSX Laboratory Refrigerators | Defrost Probe Failure: Check for loose probe connector. Replace defrost probe. |
| E03 | TSX Blood Bank Refrigerators | Defrost Probe Failure: Check for loose probe connector. Replace defrost probe. |
| E03 | TSX -30° Lab Freezers | Defrost Probe Failure: Check for loose probe connector. Replace defrost probe. |
| E03 | TSX -30° Plasma Freezers | Defrost Probe Failure: Check for loose probe connector. Replace defrost probe. |