Error code E05 on display
This article addresses error code E05, indicating an Ambient Probe Failure across various TSX models, including Laboratory Refrigerators, Blood Bank Refrigerators, -30° Lab Freezers, -30° Plasma Freezers, and -20° Laboratory and Enzyme Freezers. The resolution involves checking for a loose probe connector and replacing the ambient probe if necessary. This guidance is essential for ensuring the proper operation of these refrigeration units.
Issue
Error code E05 on display
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Lab Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | ||||
| TSX2305P | V/Y/Z | A/D | ||||
| TSX2305C | V/Y/Z | A/D | ||||
| TSX3005G | V/Y/Z | A/D | ||||
| TSX3005S | V/Y/Z | A/D | ||||
| TSX3005P | V/Y/Z | A/D | ||||
| TSX3005C | V/Y/Z | A/D | ||||
| TSX4505G | V/Y/Z | A/D | ||||
| TSX4505C | V/Y/Z | A/D | ||||
| TSX5005G | V/Y/Z | A/D | ||||
| TSX5005S | V/Y/Z | A/D | ||||
| TSX5005P | V/Y/Z | A/D | ||||
| TSX5005C | V/Y/Z | A/D |
Resolution
| Error code E05 on display | ||
| Error Code | Model Number | Resolution |
| E05 | TSX Laboratory Refrigerators | Ambient Probe Failure: Check for loose probe connector. Replace ambient probe. |
| E05 | TSX Blood Bank Refrigerators | Ambient Probe Failure: Check for loose probe connector. Replace ambient probe. |
| E05 | TSX -30° Lab Freezers | Ambient Probe Failure: Check for loose probe connector. Replace ambient probe. |
| E05 | TSX -30° Plasma Freezers | Ambient Probe Failure: Check for loose probe connector. Replace ambient probe. |
| E05 | TSX -20° Laboratory and Enzyme Freezers | Ambient Probe Failure: Check for loose probe connector. Replace ambient probe. |