Error code E06 on display
This article addresses error code E06 for TSX -30° Lab Freezers and TSX -30° Plasma Freezers, indicating a Drip Pan Probe Failure. The recommended resolution involves checking for a loose probe connector and replacing the drip pan probe if necessary. This guidance is critical for maintaining the proper functioning of these freezer models.
Issue
Error code E06 on display
Environment
| TSX -30° Lab Freezers | TSX -30° Plasma Freezers | Models | Energy Star Model |
| TSX1230F | TSX1230L | V/Y/Z | A/D |
| TSX2330F | TSX2330L | V/Y/Z | A/D |
| TSX3030F | TSX3030L | V/Y/Z | A/D |
| TSX5030F | TSX5030L | V/Y/Z | A/D |
Resolution
| Error code E06 on display | ||
| Error Code | Model Number | Resolution |
| E06 | TSX -30° Lab Freezers | Drip Pan Probe Failure: Check for loose probe connector. Replace Drip pan probe. |
| E06 | TSX -30° Plasma Freezers | Drip Pan Probe Failure: Check for loose probe connector. Replace Drip pan probe. |