Error code E07 on display
This article highlights the issue of Error E07 high compressor run time in TSX -20° Laboratory and Enzyme Freezers display. Typically problem is caused by ice buildup on the unit walls, which forces the compressor to work harder and potentially leads to thermal failure. The recommended solution is to defrost the unit to prevent further issues.
Issue
Error code E07 on display
Environment
| TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX2320F | V/Y/Z | A/D |
| TSX3020F | V/Y/Z | A/D |
| TSX2320E | V/Y/Z | A/D |
| TSX3020E | V/Y/Z | A/D |
Resolution
| Error code E07 on display | ||
| Error Code | Model Number | Resolution |
| E07 | TSX -20° Laboratory and Enzyme Freezers | Compressor Run Time High: The compressor is working harder to overcome the ice on the unit walls and may start to thermally fail. Defrost the unit. |