Location of Sensors
This article details the locations and installation methods for thermocouples in Marietta Freezers. Key thermocouples include TC6 on the heat exchanger, TC16 at the evaporator inlet, and TC18 at the evaporator outlet, installed using aluminum tape and zip ties. Additional thermocouples added in 2017 are TC2, TC12, and TC15, each with specific placement instructions to aid in troubleshooting the refrigeration system.
Issue
Thermocouple location for Marietta Freezers (Location and how to install after a repair is made)
Environment
Freezers built in Marietta starting in 2016
Resolution
Refer to the attached document for instructions.
Attachment(s)
| File | Last Modified |
|---|---|
| Location of Sensors.pdf | January 13, 2022 |