Low battery icon is lit
This article addresses the issue of a low battery icon being lit on various TSX laboratory refrigerators and freezers, including models like TSX1205G, TSX2305G, and TSX5005G. To resolve the issue, the article advises replacing the 12V backup battery located on the top right-hand side of the cabinet. The low battery icon indicates that the battery needs to be replaced.
Issue
Low battery icon is lit
Environment
| TSX Laboratory Refrigerators | TSX Blood Bank Refrigerators | TSX -30° Freezers | TSX -30° Plasma Freezers | TSX -20° Laboratory and Enzyme Freezers | Models | Energy Star Model |
| TSX1205G | TSX1204B | TSX1230F | TSX1230L | TSX2320F | V/Y/Z | A/D |
| TSX1205S | TSX2304B | TSX2330F | TSX2330L | TSX3020F | V/Y/Z | A/D |
| TSX1205P | TSX3004B | TSX3030F | TSX3030L | TSX2320E | V/Y/Z | A/D |
| TSX2305G | TSX5004B | TSX5030F | TSX5030L | TSX3020E | V/Y/Z | A/D |
| TSX2305S | V/Y/Z | A/D | ||||
| TSX2305P | V/Y/Z | A/D | ||||
| TSX2305C | V/Y/Z | A/D | ||||
| TSX3005G | V/Y/Z | A/D | ||||
| TSX3005S | V/Y/Z | A/D | ||||
| TSX3005P | V/Y/Z | A/D | ||||
| TSX3005C | V/Y/Z | A/D | ||||
| TSX4505G | V/Y/Z | A/D | ||||
| TSX4505C | V/Y/Z | A/D | ||||
| TSX5005G | V/Y/Z | A/D | ||||
| TSX5005S | V/Y/Z | A/D | ||||
| TSX5005P | V/Y/Z | A/D | ||||
| TSX5005C | V/Y/Z | A/D |
Resolution
To resolve the issue replace the battery. It is located on the top right hand side of the cabinet.
Cause
12V backup battery needs to be replaced.