Temperature validation and calibration
Issue
Unit is experiencing issues with temperature validation/calibration
Environment
TSX Ultra Low Temperature (ULT) Series:
Thermo Scientific – TSX40086A
Thermo Scientific – TSX40086D
Thermo Scientific – TSX40086V
Thermo Scientific – TSX40086G
Thermo Scientific – TSX50086A
Thermo Scientific – TSX50086D
Thermo Scientific – TSX50086V
Thermo Scientific – TSX50086G
Thermo Scientific – TSX60086A
Thermo Scientific – TSX60086D
Thermo Scientific – TSX60086V
Thermo Scientific – TSX60086G
Thermo Scientific – TSX70086A
Thermo Scientific – TSX70086D
Thermo Scientific – TSX70086V
Thermo Scientific – TSX70086G
Resolution
Possible Cause: Cabinet temperature displayed doesn’t match with actual temperature.
Solution:
- Customers performing on-site temperature calibration may observe as much as a 2°C variation when an external probe is placed next to the freezer control probe.
- This variation is normal, due to optimization of control system to ensure temperature uniformity throughout the cabinet.