TSX Gen Series - How to adjust display brightness setting
This article provides instructions on adjusting display settings for TSX Gen 2 ULT freezer series. It includes steps to adjust brightness, change the display language, set the date and time manually, change units of measure, and set the date format.
Issue:
How to change the screen brightness/intensity setting on the TSX Gen 2 ULT Freezers which includes model TSX60086A.
Environment:
TSX Gen 2 Series, TSX Gen 2 ULT Freezer Series
| TSX40086A | TSX40086D | TSX40086V |
| TSX50086A | TSX50086D | TSX50086V |
| TSX60086A | TSX60086D | TSX60086V |
| TSX70086A | TSX70086D | TSX70086V |
Resolution:
- Access the Main Menu: Press the Menu button on the device to access the main menu.
- Navigate to Settings: Use the arrow keys to scroll to the Settings option and press Enter.
- Select Display Settings: In the Settings menu, find and select Display Settings.
- Adjust Brightness/Contrast: Use the arrow keys to adjust the brightness and contrast to your preference.
- Confirm Changes: Once you have made your adjustments, press the Enter button to confirm and save the changes.
- Exit the Menu: Press the Menu button again to exit the settings menu and return to the main screen.