Unit is constantly alarming
Issue
Unit is constantly alarming
Environment
TSX Ultra Low Temperature (ULT) Series:
Thermo Scientific – TSX40086A
Thermo Scientific – TSX40086D
Thermo Scientific – TSX40086V
Thermo Scientific – TSX40086G
Thermo Scientific – TSX50086A
Thermo Scientific – TSX50086D
Thermo Scientific – TSX50086V
Thermo Scientific – TSX50086G
Thermo Scientific – TSX60086A
Thermo Scientific – TSX60086D
Thermo Scientific – TSX60086V
Thermo Scientific – TSX60086G
Thermo Scientific – TSX70086A
Thermo Scientific – TSX70086D
Thermo Scientific – TSX70086V
Thermo Scientific – TSX70086G
Resolution
Possible Causes and Solutions:
- Cause: Exterior door is closed but not sealed completely.
Solution:
- Clean any ice build-up on gasket and/or cabinet surface.
- Check for punctures or tears on gasket. Replace if necessary.
- Cause: Alarm icons appear on user interface home screen.
Solution: By pressing red bell or yellow triangle, the ticker message provides alarm details and recovery details.
- Cause: Door open alarm, exterior door not closing completely.
Solution: Open door completely and immediately close and latch it.
- Cause: Door open alarm, exterior door is closed but not sealed completely.
Solution: Defrost exterior door gasket and make sure the door is completely sealed.
- Cause: Alarm set points may have changed.
Solution: Change the set points as required.