Unit is ON but display is showing empty
Issue
Unit is ON but display is showing empty
Environment
TSX Ultra Low Temperature (ULT) Series:
Thermo Scientific – TSX40086A
Thermo Scientific – TSX40086D
Thermo Scientific – TSX40086V
Thermo Scientific – TSX40086G
Thermo Scientific – TSX50086A
Thermo Scientific – TSX50086D
Thermo Scientific – TSX50086V
Thermo Scientific – TSX50086G
Thermo Scientific – TSX60086A
Thermo Scientific – TSX60086D
Thermo Scientific – TSX60086V
Thermo Scientific – TSX60086G
Thermo Scientific – TSX70086A
Thermo Scientific – TSX70086D
Thermo Scientific – TSX70086V
Thermo Scientific – TSX70086G
Resolution
Possible Cause: User interface not powered on.
Solution:
- Push power button on user interface and hold for at least 1 second.
- Try touching the screen.
- Try restarting the unit manually by flipping the breaker at the rear of the unit.
- If this doesn’t work, call service.