Unit is over cooling
Issue
Unit is over cooling
Environment
TSX Ultra Low Temperature (ULT) Series:
Thermo Scientific – TSX40086A
Thermo Scientific – TSX40086D
Thermo Scientific – TSX40086V
Thermo Scientific – TSX40086G
Thermo Scientific – TSX50086A
Thermo Scientific – TSX50086D
Thermo Scientific – TSX50086V
Thermo Scientific – TSX50086G
Thermo Scientific – TSX60086A
Thermo Scientific – TSX60086D
Thermo Scientific – TSX60086V
Thermo Scientific – TSX60086G
Thermo Scientific – TSX70086A
Thermo Scientific – TSX70086D
Thermo Scientific – TSX70086V
Thermo Scientific – TSX70086G
Resolution
Possible Causes and Solutions:
- Cause: Set points may have changed
Solution: Adjust the set point to run at desired set point under settings.
- Cause: Temperature offset may have changed
Solution: Try adjusting the offset. Temperature offset can be set in the Controls screen under settings.
- Cause: Unknown
Solution: Try re-starting the unit. If this doesn’t help call service.