7217023 Rev 0 - Adjustable Vertical Electrophoresis System Models ADJ1_ADJ2 and ADJ3 - Operating and Maintenance Manual
The Adjustable Vertical Electrophoresis System Models ADJ1, ADJ2, and ADJ3 - Operating and Maintenance Manual" provides comprehensive guidance on the operation and maintenance of these systems. It details the specifications, safety information, setup instructions, parts list, and system assembly. The systems feature adjustable heights for the upper buffer chamber, accommodating various gel lengths. Proper use and safety precautions are emphasized to ensure optimal performance and user safety.
Description:
Operating and maintenance manual
Environment:
Adjustable Vertical Electrophoresis System, models:
- ADJ1
- ADJ2
- ADJ3
Specifications:
| Unit | ADJ-1 | ADJ-2 | ADJ-3 |
| Glass size | 16cm W x 14-40cm L | 20cm W x 20-70 cm L | 35cm W x 20-80cm L |
| Overall dimensions | 48cm H x 23cm W x 25cm D | 78cm H x 28cm W x 25cm D | 86cm H x 41 cm W x 25cm D |
| Buffer volume | 500ml | 900ml | 1800ml |
Attachment(s):
| File | Last Modified |
|---|---|
| 7217023 - Rev 0 - Adjustable Vertical Electrophoresis System, Models ADJ1, ADJ2 and ADJ3 - Operating and Maintenance Manual.pdf | December 15, 2021 |