Thermo Fisher Scientific
7217692 - Rev 0 - Gel Caster, Models JGC-3, JGC-2, and JGC-4 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual. This patented Gel Casting System is a unique method to cast 1 to 4 polyacrylamide gels without taping or special sealing of the gel plates.
Environment:
Gel Caster Models:
  • JGC-3
  • JGC-2
  • JGC-4
Specifications:
Unit/Model Number JGC-4 JGC-2 JGC-3
Gel size
10cmW x 10cmL
10cmW x 8cmL
16cmW x 14cmL
16cmW x 16cmL
20cmW x 20cmL
Footprint (cm) W x D 13 x 13 17 x 18 17 x 23
Glass Size (cm) W x L
10cmW x 10cmL
10cmW x 8cmL
16cmW x 14cmL
16cmW x 16cmL
20cmW x 20cmL
Approximate Gel Volume 6ml 15ml 28ml
Compatible with Unit P8DS, P81, P82 P9DS P10DS