Thermo Fisher Scientific
Exhaust requirements
The exhaust requirements for ovens and furnaces vary based on the process or load. Generally, they do not need an exhaust installation unless fumes need filtering, which may require a fume hood. Air flow must be assessed locally, with ovens sometimes using internal fans. Maintain specific clearances for exhaust hood and tube installations. Built-in units require air exhaust systems and should be installed in non-combustible, well-ventilated spaces.
Issue
What are the exhaust requirements for ovens and furnaces?
Environment
Ovens and furnaces
Resolution
Refer to the attached document for an answer to this question.
  • Was this article helpful?