Calibration Parameters
Issue
Resetting the LTQ Orbitrap XL ETD system tune and calibration parameters to their default values
Environment
LTQ XL Ion Trap Model
Resolution
Tips to tune and calibrate parameters:
- To reset the LTQ Orbitrap XL ETD tune and calibration parameters to their default values In the Tune Plus window.
- Choose File > Restore Factory Calibration to restore the default calibration parameters.
OR
- Choose File > Restore Factory Tune Method to restore the default tune parameters.
Note: Make sure that any problems you might be experiencing are not due to improper API source settings (spray voltage, sheath and auxiliary gas flow, ion transfer capillary temperature, etc.) before resetting the system parameters to their default values.