Thermo Fisher Scientific
Peak Shape and Recovery Problems for Polar Opiates
Issue
Instrument experiencing poor recoveries as some of the analyte molecules eluted as “breakthrough” peaks far ahead of their expected retention times.
Environment
Tribrid MS
Resolution
1. Change starting gradient conditions from 5 or 10% methanol to 100% aqueous mobile phase. Figure 1 shows how the elution and shapes of polar opiates changed by this step.
clipboard_ea0016a1959f06ad3124ee97c9c0edadf.png
Figure 1: Gradients start with 95% aqueous + 5% methanol
2. Ensure completed column equilibration with 100% aqueous mobile phase. Figures 2 and 3 show column pressure traces typical of insufficient and sufficient equilibration.
clipboard_eefe623b859c4f634ec4661e3628b44a3.png
Figure 2: PFP column pressure trace showing insufficient equilibration
clipboard_e2b7c81115cf832a4bb11787d2ebacee3.png
Figure 3: PFP column pressure trace showing sufficient equilibration
3. Change autosampler method by introducing an air gap between washes and samples.
4. Ensure that no residual Wash 2 remained in injection ports after washes. Excessive needle gaps during rinses with organic wash caused peak problems.