Signal Issues
Issue
The signal intensity is weak.
Environment
Easy Spray Ion Spray Sources Models:
- ES081
- ES082
Resolution
- Verify that the EASY-Spray column is correctly positioned (that is, clicks on insertion).
- Check the LC method and MS tune method parameters.
- Run a known standard to check the sensitivity.
- Clean the ion transfer tube.
Cause
- The parameters for the LC, MS, or both might need adjusting.
- The MS ion transfer tube is dirty.