Thermo Fisher Scientific
Replacing the TurboFlow Column every 300-1000 injections
Issue
How do I replace the TurboFlow column?
NOTE: Use the Maintenance dialog box to track injection numbers.
Environment
  • LX-2
  • LX-4
  • TLX-1
  • TLX-2
  • TLX-4
Resolution
1. Turn off the column heater and allow the column heater to cool to room temperature.
       a. Open the Aria MX Direct Control window.
       b. Click a column heater that appears under the appropriate channel name.
       c. To turn off the column heater, click the Enable button so that it shows dark green.
       d. Repeat step b and step c to turn off the second column heater.
2. When the column heater has cooled, unwrap the column heater from the column.
3. Replace the column as directed.
Note: Use only your fingers to manipulate the tubing fittings on Prelude systems. Transcend systems require tools, depending on the procedure.